2,3,5,6-Tetrachloroaniline

Supplier: EHRENSTORFER GMBH

EHERC17330600EA 39.7 EUR
EHERC17330600
2,3,5,6-Tetrachloroaniline
2,3,5,6-Tetrachloroaniline
Formula: C₆H₃Cl₄N
MW: 230,91 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 110 °C
MDL Number: MFCD00007659
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR