2,3-Dichloroaniline ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
A11524.22
A11524.36
A11524.22EA
79.7
EUR
A11524.22
A11524.36
2,3-Dichloroaniline ≥99%
2,3-Dichloroaniline
Purity: 99%
CAS No.: 608-27-5
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 115°(239°F)
Refractive Index: 1.6060
Melting Point: 20-24°
Boiling Point: 250-252°
Beilstein Reference: 472027
UN No.: 3442
MDL No.: MFCD00007657
Formula:
C₆H₅Cl₂N Boiling Pt: 250…252 °C Melting Pt: 20…24 °C Density: 1.37 Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007657 CAS Number: 608-27-5 EINECS: 210-157-9 UN: 3442 ADR: 6.1,II |