2,4,6-Trichloroaniline
Supplier: Thermo Scientific
Danger
SB01076.1 SB01076.10 SB01076.50
MAYBSB01076.1EA 0 EUR
MAYBSB01076.1 MAYBSB01076.10 MAYBSB01076.50
2,4,6-Trichloroaniline
2,4,6-Trichloroaniline
Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
Boiling Pt: 262
Melting Pt: 74
Storage Temperature: Ambient
MDL Number: MFCD00007663
CAS Number: 634-93-5
EINECS: 211-219-8
UN: 2018
ADR: 6.1 II