3,5-Dichloroaniline ≥98%
Supplier: Thermo Scientific
Danger
A13922.0B A13922.18 A13922.30
A13922.0BEA 401 EUR
A13922.0B A13922.18 A13922.30
3,5-Dichloroaniline ≥98%
3,5-Dichloroaniline
Purity: 98% CAS No.: 626-43-7 Molecular Formula: C6H5Cl2N Formula Weight: 162.02 Flash Point: 133°(271°F) Melting Point: 49-53° Boiling Point: 259-260°/741mm Beilstein Reference: 636492 UN No.: 3442 MDL No.: MFCD00007774
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 259…260 °C (741 mmHg)
Melting Pt: 49…53 °C
Density: 1.58
Flash Pt: 133 °C (271 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II