3,5-Dichloroaniline
Supplier: Thermo Scientific
|
Danger
|
SB00590.1
SB00590.10
SB00590.50
MAYBSB00590.1EA
0
EUR
MAYBSB00590.1
MAYBSB00590.10
MAYBSB00590.50
3,5-Dichloroaniline
3,5-Dichloroaniline
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 262 °C (1013 hPa) Melting Pt: 51 °C Density: 1.58 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007774 CAS Number: 626-43-7 EINECS: 210-948-9 UN: 3442 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...