Laboratory bottles, narrow neck, with screw cap and thread
Supplier: Lenz Laborglas GmbH & CO.KG
07105022 07105028 07105037 07105049 07105058 07105070 07105073 07105082 07105088 07105091 07105094
LENZ07105022EA 124 EUR
LENZ07105022 LENZ07105028 LENZ07105037 LENZ07105049 LENZ07105058 LENZ07105070 LENZ07105073 LENZ07105082 LENZ07105088 LENZ07105091 LENZ07105094
Laboratory bottles, narrow neck, with screw cap and thread
Bottles Glass Bottles
Bottles are made of DURAN® borosilicate glass, with DIN thread, blue PP screw caps and pouring rings.
  • Studded base for stability
  • Resistant to abrasions
  • With graduation and inscription label
Certifikace: ISO 4796