2,4,5-Trichlorophenol

Supplier: EHRENSTORFER GMBH

EHERC17774500EA 55.8 EUR
EHERC17774500
2,4,5-Trichlorophenol
2,4,5-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197,45 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 68 °C
Density: 1,68 g/cm³ (20 °C)
MDL Number: MFCD00002170
UN: 3077
ADR: 9,III
Merck Index: 12,09772

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR