2,4-Dichlorothiophenol
Supplier: Thermo Scientific
Warning
Synonyms: 2,4-Dichlorobenzenethiol, 1,3-Dichloro-4-mercaptobenzene, 2,4-Dichlorobenzene-1-thiol
TL00291.10 TL00291.50
MAYBTL00291.10EA 0 EUR
MAYBTL00291.10 MAYBTL00291.50
2,4-Dichlorothiophenol
2,4-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 115 °C (8 mmHg)
Melting Pt: 22
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7