1,2-Difluorobenzene
Supplier: Thermo Scientific
SB00649.1 SB00649EA SB00649.50
MAYBSB00649.1EA 0 EUR
MAYBSB00649.1 MAYBSB00649EA MAYBSB00649.50
1,2-Difluorobenzene
1,2-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000284
CAS Number: 367-11-3
EINECS: 206-680-7
UN: 1993
ADR: 3,II