Laboratory knives
Supplier: RSG
199.001 199.002 199.003 199.004
233-0146EA 105 EUR
233-0146 233-0147 233-0148 233-0149
Laboratory knives
Knives
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
  • Anti-magnetic and lightweight
  • Dishwasher-proof
  • Long-life sharpness