1,4-Difluorobenzene
Supplier: Thermo Scientific
SB01462.1
SB01462.10
SB01462.50
MAYBSB01462.1EA
0
EUR
MAYBSB01462.1
MAYBSB01462.10
MAYBSB01462.50
1,4-Difluorobenzene
1,4-Difluorobenzene
Formula:
C₆H₄F₂ MW: 114.09 g/mol Boiling Pt: 88 °C (1013 hPa) Melting Pt: –13 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000344 CAS Number: 540-36-3 EINECS: 208-742-9 UN: 1993 ADR: 3,II Merck Index: 13,03172 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...