2,4,6-Trifluorobenzonitrile
Supplier: Thermo Scientific
SB01909.1
SB01909.10
MAYBSB01909.1EA
0
EUR
MAYBSB01909.1
MAYBSB01909.10
2,4,6-Trifluorobenzonitrile
2,4,6-Trifluorobenzonitrile
Formula:
C₇H₂F₃N Boiling Pt: 179 °C (1013 hPa) Melting Pt: 57…58 °C |
MDL Number:
MFCD00042399 CAS Number: 96606-37-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...