2,6-Difluorobenzonitrile
Supplier: Thermo Scientific
|
Warning
|
SB00299.1
SB00299.10
SB00299.50
MAYBSB00299.1EA
0
EUR
MAYBSB00299.1
MAYBSB00299.10
MAYBSB00299.50
2,6-Difluorobenzonitrile
2,6-Difluorobenzonitrile
Formula:
C₇H₃F₂N MW: 139.1 g/mol Boiling Pt: 98…100 °C (31 mmHg) Melting Pt: 30…32 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001778 CAS Number: 1897-52-5 EINECS: 217-589-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...