2-Fluorobenzonitrile 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: o-Fluorobenzonitrile
250050250 250050050 250051000
ACRO250050250EA 74.8 EUR
ACRO250050250 ACRO250050050 ACRO250051000
2-Fluorobenzonitrile 99%
2-Fluorobenzonitrile
Formula: C₇H₄FN
Boiling Pt: 90 °C (21 mmHg)
Melting Pt: 35…37 °C
Density: 1.11 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS Number: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5063 to 1.5083 (20°C, 589 nm)
GC ≥98.5 %