2-Fluorobenzonitrile
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
o-Fluorobenzonitrile
SB00740.1
SB00740.10
SB00740.50
MAYBSB00740.1EA
0
EUR
MAYBSB00740.1
MAYBSB00740.10
MAYBSB00740.50
2-Fluorobenzonitrile
2-Fluorobenzonitrile
Formula:
C₇H₄FN Boiling Pt: 190 °C (1013 hPa) Melting Pt: 35…37 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001773 CAS Number: 394-47-8 EINECS: 206-897-7 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...