2,4-Difluorobenzoic acid
Supplier: Thermo Scientific
|
Warning
|
TL00169.1
TL00169.10
TL00169.50
MAYBTL00169.1EA
0
EUR
MAYBTL00169.1
MAYBTL00169.10
MAYBTL00169.50
2,4-Difluorobenzoic acid
2,4-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ Boiling Pt: 241 °C (1013 hPa) Melting Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011670 CAS Number: 1583-58-0 EINECS: 216-430-9 |