2,6-Difluorobenzoic acid
Supplier: Thermo Scientific
Warning
SB01105DA SB01105EA SB01105.50
MAYBSB01105DAEA 0 EUR
MAYBSB01105DA MAYBSB01105EA MAYBSB01105.50
2,6-Difluorobenzoic acid
2,6-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
Boiling Pt: 72…77 °C (13 mmHg)
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002411
CAS Number: 385-00-2
EINECS: 206-856-3