2-Fluorobenzoic acid
Supplier: Thermo Scientific
Warning
SB00741.250 SB00741.1
MAYBSB00741.250EA 0 EUR
MAYBSB00741.250 MAYBSB00741.1
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 121…124 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1