3,4-Difluorobenzoic acid
Supplier: Thermo Scientific
|
Warning
|
SB01146.1
SB01146.10
SB01146.50
MAYBSB01146.1EA
0
EUR
MAYBSB01146.1
MAYBSB01146.10
MAYBSB01146.50
3,4-Difluorobenzoic acid
3,4-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 119…122 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011672 CAS Number: 455-86-7 EINECS: 207-249-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...