4-Fluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Warning
A14129.14 A14129.22 A14129.36
A14129.14EA 37.6 EUR
A14129.14 A14129.22 A14129.36
4-Fluorobenzoic acid ≥98%
4-Fluorobenzoic acid
Purity: 98% CAS No.: 456-22-4 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 183-187° Beilstein Reference: 1906922 Merck Reference: 14,4171 MDL No.: MFCD00002530
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 254
Melting Pt: 183…187 °C
Density: 1.470
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 13,04198