Pentafluorobenzoic acid ≥99%
Supplier: Thermo Scientific
Warning
Synonyms: 2,3,4,5,6-Pentafluorobenzoic acid
A12396.06 A12396.14
A12396.06EA 29.6 EUR
A12396.06 A12396.14
Pentafluorobenzoic acid ≥99%
Pentafluorobenzoic acid
Purity: 99% CAS No.: 602-94-8 Molecular Formula: C7HF5O2 Formula Weight: 212.07 Melting Point: 100-104° Boiling Point: 219-221° MDL No.: MFCD00002406
Formula: C₇HF₅O₂
MW: 212.08 g/mol
Boiling Pt: 219…221 °C
Melting Pt: 100…104 °C
Density: 1.942 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002406
CAS Number: 602-94-8
EINECS: 210-026-6