6-Fluoroanthranilic acid

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-6-fluorobenzoic acid

BTB07342.1 BTB07342.10
MAYBBTB07342.1EA 0 EUR
MAYBBTB07342.1 MAYBBTB07342.10
6-Fluoroanthranilic acid
6-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 305
Melting Pt: 165…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00067781
CAS Number: 434-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR