4-(Trifluoromethoxy)benzoic acid

Supplier: Thermo Scientific
Warning

TL00376.1 TL00376.10
MAYBTL00376.1EA 0 EUR
MAYBTL00376.1 MAYBTL00376.10
4-(Trifluoromethoxy)benzoic acid
4-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 153,4 °C
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR