2-Fluoro-6-iodobenzoic acid ≥97%
Supplier: Thermo Fisher Scientific
Warning
L07924.03 L07924.06
L07924.03EA 33.6 EUR
L07924.03 L07924.06
2-Fluoro-6-iodobenzoic acid ≥97%
2-Fluoro-6-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Boiling Pt: 312
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00042289
CAS Number: 111771-08-5