2,3,4-Trifluoroaniline
Supplier: Thermo Scientific
|
Danger
|
SB01064.1
SB01064.10
SB01064.50
MAYBSB01064.1EA
0
EUR
MAYBSB01064.1
MAYBSB01064.10
MAYBSB01064.50
2,3,4-Trifluoroaniline
2,3,4-Trifluoroaniline
Formula:
C₆H₄F₃N MW: 147.1 g/mol Boiling Pt: 174 Melting Pt: 14…15 °C Density: 1.39 g/cm³ (20 °C) |
MDL Number:
MFCD00011737 CAS Number: 3862-73-5 EINECS: 407-170-9 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...