2,5-Difluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A10272.09
A10272.18
A10272.09EA
47.1
EUR
A10272.09
A10272.18
2,5-Difluoroaniline ≥98%
2,5-Difluoroaniline
Purity: 98%
CAS No.: 367-30-6
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Flash Point: 68°(154°F)
Refractive Index: 1.5130
Odour: Characteristic
Melting Point: 13-14°
Boiling Point: 171-174°
Beilstein Reference: 2802549
UN No.: 2941
MDL No.: MFCD00007651
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 171…174 °C Melting Pt: 13…14 °C Density: 1.285 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007651 CAS Number: 367-30-6 EINECS: 206-690-1 UN: 2941 ADR: 6.1 III |