3,5-Difluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Warning
A16528.06 A16528.14 A16528.22
A16528.06EA 52 EUR
A16528.06 A16528.14 A16528.22
3,5-Difluoroaniline ≥98%
3,5-Difluoroaniline
Purity: 98+% CAS No.: 372-39-4 Molecular Formula: C6H5F2N Formula Weight: 129.11 Flash Point: 75°(167°F) Odour: Characteristic Boiling Point: 80°/20mm Beilstein Reference: 2802286 UN No.: 2811 MDL No.: MFCD00007763
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 80 °C (20 mmHg)
Melting Pt: 37…41 °C
Density: 1.295
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1 III