4-Fluoroaniline 98%
Supplier: Thermo Fisher Scientific
Danger
119291000 119295000 119290250
ACRO119291000EA 50.2 EUR
ACRO119291000 ACRO119295000 ACRO119290250
4-Fluoroaniline 98%
4-Fluoroaniline
Formula: C₆H₆FN
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –1.9 °C
Density: 1.157 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196
Order Now
Specification Test Results
Appearance Clear light yellow to red-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20°C, 589 nm)
GC ≥97.5 %