4-Fluoroaniline, Sigma-Aldrich®
Supplier: Merck
Warning
F3800-25G F3800-100G F3800-500G
SIALF3800-25GEA 22.7 EUR
SIALF3800-25G SIALF3800-100G SIALF3800-500G
4-Fluoroaniline, Sigma-Aldrich®
4-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196