4-Fluoro-2-nitroaniline
Supplier: Thermo Scientific
BTB02200.1 BTB02200.10
MAYBBTB02200.1EA 0 EUR
MAYBBTB02200.1 MAYBBTB02200.10
4-Fluoro-2-nitroaniline
4-Fluoro-2-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 95
Storage Temperature: Ambient
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0