4-Fluoro-3-nitroaniline 99%
Supplier: Thermo Scientific
Warning
168400250 168401000 168400050
ACRO168400250EA 46 EUR
ACRO168400250 ACRO168401000 ACRO168400050
4-Fluoro-3-nitroaniline 99%
4-Fluoro-3-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 96…99 °C
Density: 0,43 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5
Order Now
Specification Test Results
Appearance Ochre Powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
GC ≥98.5 %