3,4-Difluoroanisole ≥98%
Supplier: Thermo Scientific
Warning
A12474.06 A12474.14 A12474.22
A12474.06EA 29.8 EUR
A12474.06 A12474.14 A12474.22
3,4-Difluoroanisole ≥98%
3,4-Difluoroanisole
Purity: 98% CAS No.: 115144-40-6 Molecular Formula: C7H6F2O Formula Weight: 144.12 Flash Point: 55°(131°F) Refractive Index: 1.4680 Beilstein Reference: 6717610 UN No.: 1993 MDL No.: MFCD00010730
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.214
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010730
CAS Number: 115144-40-6
UN: 1993
ADR: 3,III