Pentafluoroanisole ≥98%

Supplier: Thermo Fisher Scientific
Warning

B23928.06 B23928.14 B23928.22
B23928.06EA 34.1 EUR
B23928.06 B23928.14 B23928.22
Pentafluoroanisole ≥98%
Pentafluoroanisole

Purity: 98% CAS No.: 389-40-2 Molecular Formula: C7H3F5O Formula Weight: 198.09 Flash Point: 32°(89°F) Refractive Index: 1.4090 Melting Point: -38° Boiling Point: 136-138° Beilstein Reference: 2052664 UN No.: 1993 MDL No.: MFCD00000289

Formula: C₇H₃F₅O
Boiling Pt: 136…138 °C
Melting Pt: –38 °C
Density: 1.50
Flash Pt: 32 °C (89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000289
CAS Number: 389-40-2
EINECS: 206-866-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR