2-Fluorophenol

Supplier: Thermo Scientific
Danger

SB00849.1 SB00849.10 SB00849.50
MAYBSB00849.1EA 0 EUR
MAYBSB00849.1 MAYBSB00849.10 MAYBSB00849.50
2-Fluorophenol
2-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR