5-Fluoro-2-nitrophenol

Supplier: Thermo Scientific
Warning

Synonyms: 4-Fluoro-2-hydroxynitrobenzene

SB00633.1 SB00633.10 SB00633.50
MAYBSB00633.1EA 0 EUR
MAYBSB00633.1 MAYBSB00633.10 MAYBSB00633.50
5-Fluoro-2-nitrophenol
5-Fluoro-2-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6
EINECS: 207-168-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR