3-Fluorophenol
Supplier: Thermo Scientific
Warning
SB01944.1 SB01944.10 SB01944.50
MAYBSB01944.1EA 0 EUR
MAYBSB01944.1 MAYBSB01944.10 MAYBSB01944.50
3-Fluorophenol
3-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 13.7 °C
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,II