3,4,5-Trifluorophenol, Sigma-Aldrich®
Supplier: Merck
51092-1G-F
51092-5G-F
SIAL51092-1G-FEA
25
EUR
SIAL51092-1G-F
SIAL51092-5G-F
3,4,5-Trifluorophenol, Sigma-Aldrich®
3,4,5-Trifluorophenol
Formula:
C₆H₃F₃O MW: 148.08 g/mol Density: 1.629 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042427 CAS Number: 99627-05-1 EINECS: 436-120-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...