4-Fluorothiophenol
Supplier: Thermo Scientific
Danger
Synonyms: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol
SB01479.1 SB01479.10 SB01479.50
MAYBSB01479.1EA 0 EUR
MAYBSB01479.1 MAYBSB01479.10 MAYBSB01479.50
4-Fluorothiophenol
4-Fluorothiophenol
Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 164
Density: 1.1996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004846
CAS Number: 371-42-6
EINECS: 206-737-6
UN: 3071
ADR: 6.1 II