2-Iodobenzoic acid
Supplier: Thermo Scientific
Synonyms: o-Iodobenzoic acid
BTB05694.250 BTB05694.1
MAYBBTB05694.250EA 0 EUR
MAYBBTB05694.250 MAYBBTB05694.1
2-Iodobenzoic acid
2-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 314
Melting Pt: 159…160 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052