1-Iodo-4-nitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Warning
B23950.06 B23950.14 B23950.22
B23950.06EA 33.4 EUR
B23950.06 B23950.14 B23950.22
1-Iodo-4-nitrobenzene ≥98%
1-Iodo-4-nitrobenzene
Purity: 98+% CAS No.: 636-98-6 Molecular Formula: C6H4INO2 Formula Weight: 249.01 Melting Point: 172-177° Boiling Point: 288-290° MDL No.: MFCD00007299
Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 288…290 °C
Melting Pt: 172…177 °C
Density: 2.273 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7