3-Iodophenol ≥98%
Supplier: Thermo Fisher Scientific
Warning
A11155.09 A11155.18
A11155.09EA 59.5 EUR
A11155.09 A11155.18
3-Iodophenol ≥98%
3-Iodophenol
Purity: 98% CAS No.: 626-02-8 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 38-43° Beilstein Reference: 2039304 MDL No.: MFCD00002261
Formula: C₆H₅IO
MW: 220.01 g/mol
Melting Pt: 38…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2
UN: 3335
ADR: 9