Industrial scales, Excellence XA series
Supplier: Mettler - Toledo
11126840 11126845
611-2480EA 0 EUR
611-2480 611-2481
Industrial scales, Excellence XA series
Balances and Scales
The large pan and high capacity versions of the XA series in the Excellence range are simple to use and provide fast, reliable, accurate weighing using MonoBloc HighSpeed technology. The monochrome touch screen provides fast and easy access to built-in applications.
  • Durable, robust design with large stainless steel pan
  • FACT; fully automatic temperature controlled internal adjustment
  • Chemically resistant housing and overload protection for everyday use in difficult weighing environments
  • Intuitive interface with data input via alphanumeric touchpad
  • Easy to