Semi-micro and analytical balances, NewClassic series
Supplier: Mettler - Toledo
11124782 11124479 11124474 11124477 11124480 11124770 11124776 11142062 11142056 11142068 11142074
611-2469EA 0 EUR
611-2469 METB11124479 611-2464 611-2465 611-2466 611-2467 611-2468 611-2471 611-2470 611-2472 611-2473
Semi-micro and analytical balances, NewClassic series
Balances and Scales
A stylish series of traditional analytical balances for simple weighing tasks. The balances all include the following benefits with more specific features listed for the three ranges below:
  • SmartTrac; shows how much of the balance range has been used
  • Balance diagnostics
  • Protection of balance configurations against manipulation and also overload protection
Semi-micro NewClassic MS balances feature:
HRT (HiRes Technology) weighing cell
FACT fully automatic time and temperature controlled internal adjustment
ErgoDoor; the right door of the draught shieldMore Product Information