Microbalance, Excellence XS3DU
Supplier: Mettler-Toledo
611-2454EA 0 EUR
611-2454
Microbalance, Excellence XS3DU
Balances and Scales
An economical microbalance that delivers reliable measurement performance for straightforward weighing of microgramme samples.
  • Monochrome touch screen display for simple intuitive control
  • FACT fully automatic time and temperature controlled internal adjustment
  • SmartTrac to provide a graphical track of capacity and weighing tolerances
  • Manually driven draught shield
  • Slot for second interface to provide range of connectivity options
The five pre-programmed applications and three user-definable fields for sample and operator identification provide sufficient data for simple weighing tasks.
Informace pro objednávání: For details of the full range of accessories, including options for filter weighing and antistatic options, please contact your local Avantor sales office.
Upozornení: * Measured at low load conditions (0,2g)