N,N-Dimethyl-4-nitrosoaniline ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Nitroso-N,N-dimethylaniline
A18301.14 A18301.22 A18301.30
A18301.14EA 36.4 EUR
A18301.14 A18301.22 A18301.30
N,N-Dimethyl-4-nitrosoaniline ≥98%
N,N-Dimethyl-4-nitrosoaniline
Purity: 98% CAS No.: 138-89-6 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 84-87° Beilstein Reference: 607293 Merck Reference: 14,6638 UN No.: 1369 MDL No.: MFCD00002063
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 259
Melting Pt: 84…87 °C
Density: 1.150
Storage Temperature: Ambient
MDL Number: MFCD00002063
CAS Number: 138-89-6
EINECS: 205-343-1
UN: 1369
ADR: 4.2 II
Merck Index: 12,06736