3,5-Dimethoxyaniline

Supplier: Thermo Scientific
Warning

SB00908.1 SB00908.10 SB00908.50
MAYBSB00908.1EA 0 EUR
MAYBSB00908.1 MAYBSB00908.10 MAYBSB00908.50
3,5-Dimethoxyaniline
3,5-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR