2-Nitrophenol 99%
Supplier: Thermo Scientific
Warning
Synonyms: o-Nitrophenol
128740010 128740050 128741000 128745000
ACRO128740010EA 115 EUR
ACRO128740010 ACRO128740050 ACRO128741000 ACRO128745000
2-Nitrophenol 99%
2-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 214.6 °C (1013 hPa)
Melting Pt: 43…47 °C
Density: 1,599 g/cm³ (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1,III
Merck Index: 12,06717
Order Now
Specification Test Results
Appearance Yellow to yellow-green Powder or crystals and/or chunks or low melting crystalline mass
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥98.5 %