Biohazard Transport Carriers
Supplier: Kartell
plastilab®
216-4305EA 260 EUR
216-4305
Biohazard Transport Carriers
Carriers Test Tubes Transport Carriers
Transparent PC, silicone gasket and stainless steel handle.
  • Completely autoclavable
  • Biohazard symbol on the box
  • User sheet with instructions approved by WHO
To carry tubes or urine stool containers under safe conditions.
  • Completely autoclavable
  • Biohazard symbol on the box
  • User sheet with instructions approved by WHO
To carry tubes or urine stool containers under safe conditions.