2,3-Dinitrophenol stabilised, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Stabilization:
Moistened with water
42155-1G
42155-5G
SIAL42155-1GEA
383
EUR
SIAL42155-1G
SIAL42155-5G
2,3-Dinitrophenol stabilised, Sigma-Aldrich®
2,3-Dinitrophenol
Formula:
C₆H₄N₂O₅ MW: 184.11 g/mol Melting Pt: 144 °C Density: 1.681 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
66-56-8 EINECS: 200-628-7 UN: 1320 ADR: 4.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...