Guaiacol
Supplier: Cayman Chemical
CAYM70430-25EA 16 EUR
CAYM70430-25 CAYM70430-500 CAYM70430-100 CAYM70430-50
Guaiacol
Guaiacol
Formula: C₇H₈O₂
MW: 124,14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1,1534 g/cm³ (0 °C)
Flash Pt: 82 °C
MDL Number: MFCD00002185
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568